siRNA Details
| Gene Symbol: C1qtnf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190313.1 | NP_001177242.1 | 6 | 851 | ||
| NM_145613.4 | NP_663588.2 | 6 | 851 | |||
| GenBank mRNA | AF469650.1 | - | 6 | 779 | ||
| AK053629.1 | - | 6 | 852 | |||
| Gene Symbol: Mfrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190314.1 | NP_001177243.1 | 6 | 851 | ||
| NM_147126.3 | NP_667337.1 | 6 | 851 | |||

C1qtnf5
Genomic Map