siRNA Details
| Gene Symbol: Ehd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153068.3 | NP_694708.2 | 6 | 1524 | ||
| GenBank mRNA | AK029240.1 | - | 6 | 1469 | ||
| AK031203.1 | - | 6 | 1526 | |||
| AK046566.1 | - | 6 | 1466 | |||
| AK048356.1 | - | 6 | 1469 | |||
| AK050243.1 | - | 6 | 1504 | |||
| AK084455.1 | - | 6 | 1467 | |||
| AK085016.1 | - | 6 | 1467 | |||
| AY531389.1 | - | 6 | 1463 | |||
| BC027084.1 | - | 7 | 1171 | 5356224 | ||
| BC031731.1 | - | 1389 | ||||
| BC113161.1 | - | 7 | 1482 | 40060184 | ||

Ehd2
Genomic Map