siRNA Details
| Gene Symbol: PSMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191037.1 | NP_001177966.1 | 9 | 1185 | ||
| NM_002807.3 | NP_002798.2 | 9 | 1185 | |||
| NR_034059.1 | 8 | 1015 | ||||
| XM_017004517.1 | XP_016860006.1 | 9 | 1185 | |||
| GenBank mRNA | AK307415.1 | - | 8 | 1012 | ||
| AK307513.1 | - | 7 | 887 | |||
| AK312528.1 | - | 9 | 1142 | |||
| BC001053.2 | - | 9 | 1142 | |||
| BC014013.2 | - | 9 | 1154 | |||
| BC020223.1 | - | 9 | 1132 | |||
| BC032445.1 | - | 9 | 1157 | |||
| BC039845.1 | - | 9 | 1108 | |||
| BC047897.1 | - | 9 | 1107 | |||
| BC064398.1 | - | 9 | 1156 | |||
| BC073833.1 | - | 9 | 1109 | |||
| BC094720.1 | - | 9 | 1171 | 4826340 | ||
| BC112344.1 | - | 9 | 1146 | 40008454 | ||
| BC114434.1 | - | 9 | 1146 | 40008444 | ||
| D44466.1 | - | 9 | 1103 | |||
| KU921434.1 | - | 2 | 142 | |||

PSMD1
Genomic Map