siRNA Details
| Gene Symbol: Gga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252067.1 | NP_001238996.1 | 8, 9 | 804 | ||
| NM_173048.3 | NP_766636.2 | 8, 9 | 804 | |||
| GenBank mRNA | AK031086.1 | - | 8, 9 | 776 | ||
| AK082327.1 | - | 8, 9 | 768 | |||
| AK122212.2 | - | 8, 9 | 773 | |||
| AK132183.1 | - | 8, 9 | 772 | |||
| AK147882.1 | - | 8, 9 | 773 | |||
| AK213811.1 | - | 4, 5 | 367 | |||
| BC024266.1 | - | 1411 | ||||
| BC056424.1 | - | 8, 9 | 758 | |||
| BC110676.1 | - | 8, 9 | 760 | |||

Gga3
Genomic Map