siRNA Details
| Gene Symbol: Nphp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153424.2 | NP_700473.2 | 13, 14 | 1964 | ||
| GenBank mRNA | AK038879.1 | - | 13, 14 | 1719 | ||
| AK084539.1 | - | 13, 14 | 1966 | |||
| AK172998.1 | - | 10, 11 | 1277 | |||
| AY118229.1 | - | 12, 13 | 1656 | |||
| BC138370.1 | - | 13, 14 | 1689 | 8861390 | ||

Nphp4
Genomic Map