siRNA Details
| Gene Symbol: Nphp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153424.2 | NP_700473.2 | 5, 6 | 875 | ||
| GenBank mRNA | AF537131.1 | - | 5, 6 | 829 | ||
| AK038879.1 | - | 5, 6 | 630 | |||
| AK084539.1 | - | 5, 6 | 875 | |||
| AK172998.1 | - | 2, 3 | 188 | |||
| AY118229.1 | - | 4, 5 | 567 | |||
| BC138370.1 | - | 5, 6 | 600 | 8861390 | ||

Nphp4
Genomic Map