siRNA Details
| Gene Symbol: Cdk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153599.3 | NP_705827.2 | 2, 3 | 228 | ||
| GenBank mRNA | AK131948.1 | - | 2, 3 | 168 | ||
| BI151839.1 | - | 2, 3 | 228 | |||
| CF730982.1 | - | 2, 3 | 146 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk8| Gene Symbol: Cdk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153599.3 | NP_705827.2 | 2, 3 | 228 | ||
| GenBank mRNA | AK131948.1 | - | 2, 3 | 168 | ||
| BI151839.1 | - | 2, 3 | 228 | |||
| CF730982.1 | - | 2, 3 | 146 | |||