siRNA Details
| Gene Symbol: Ttc26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153600.2 | NP_705828.2 | 7 | 731 | ||
| GenBank mRNA | AK035185.1 | - | 7 | 739 | ||
| BC066060.1 | - | 7 | 731 | 6848602 | ||
| BC066087.1 | - | 8 | 921 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc26| Gene Symbol: Ttc26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153600.2 | NP_705828.2 | 7 | 731 | ||
| GenBank mRNA | AK035185.1 | - | 7 | 739 | ||
| BC066060.1 | - | 7 | 731 | 6848602 | ||
| BC066087.1 | - | 8 | 921 | |||