siRNA Details
| Gene Symbol: Acsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153807.2 | NP_722502.1 | 9 | 1097 | ||
| GenBank mRNA | AK034209.1 | - | 9 | 1122 | ||
| AK153582.1 | - | 9 | 1129 | |||
| AK199766.1 | - | 4 | 334 | |||
| BC018371.1 | - | 9 | 1097 | 4210220 | ||
| BC063269.1 | - | 9 | 1182 | 4159843 | ||

Acsf2
Genomic Map