siRNA Details
Gene Symbol: BB287469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177573.1 | NP_001171044.1 | 2 | 1403 | ||
GenBank mRNA | AK139752.1 | - | 2 | 1403 |
Gene Symbol: Gm5039 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094938.1 | - | 1 | 1188 |
