siRNA Details
| Gene Symbol: Irak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029926.5 | NP_084202.2 | 2 | 322 | ||
| GenBank mRNA | AF445803.1 | - | 2 | 310 | ||
| AK020397.2 | - | 2 | 330 | |||
| AK028837.1 | - | 2 | 263 | |||
| AK029028.1 | - | 2 | 323 | |||
| AK139992.1 | - | 311 | ||||
| BC051676.1 | - | 2 | 315 | 30017484 | ||

Irak4
Genomic Map