siRNA Details
| Gene Symbol: Lgsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153601.1 | NP_705829.1 | 5 | 1653 | ||
| GenBank mRNA | AF545851.1 | - | 5 | 1653 | ||
| BC117745.1 | - | 5 | 1653 | 40090427 | ||
| BC117746.1 | - | 5 | 1653 | 40090429 | ||
| BC128067.1 | - | 5 | 1653 | 40090416 | ||

Lgsn
Genomic Map