siRNA Details
| Gene Symbol: Samd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013766.2 | NP_001013788.2 | 7 | 844 | ||
| NM_001115154.1 | NP_001108626.1 | 7 | 844 | |||
| GenBank mRNA | AK082201.1 | - | 7 | 845 | ||
| AK144462.1 | - | 4 | 681 | |||
| BC150876.1 | - | 7 | 845 | 9087787 | ||

Samd3
Genomic Map