siRNA Details
| Gene Symbol: Rnf217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146349.1 | NP_001139821.1 | 2 | 1415 | ||
| GenBank mRNA | BF467827.1 | - | 2 | 200 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf217| Gene Symbol: Rnf217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146349.1 | NP_001139821.1 | 2 | 1415 | ||
| GenBank mRNA | BF467827.1 | - | 2 | 200 | ||