siRNA Details
| Gene Symbol: Scml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172938.3 | NP_766526.3 | 4, 5 | 772 | ||
| GenBank mRNA | AF397911.1 | - | 4, 5 | 640 | ||
| AK034171.1 | - | 4, 5 | 611 | |||
| AK047972.1 | - | 2, 3 | 198 | |||
| AK220309.1 | - | 2, 3 | 156 | |||
| BC043310.1 | - | 4, 5 | 567 | 5067233 | ||

Scml4
Genomic Map