siRNA Details
| Gene Symbol: Scml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172938.3 | NP_766526.3 | 9 | 1621 | ||
| GenBank mRNA | AF397912.1 | - | 2 | 158 | ||
| AK034171.1 | - | 9 | 1460 | |||
| AK040597.1 | - | 4 | 1269 | |||
| AK043500.1 | - | 4 | 1152 | |||
| AK047972.1 | - | 7 | 1047 | |||
| AK141826.1 | - | 4 | 645 | |||
| AK172160.1 | - | 5 | 684 | |||
| AK220309.1 | - | 7 | 1005 | |||
| BC024570.1 | - | 4 | 577 | |||
| BC039567.1 | - | 2 | 144 | |||
| BC043310.1 | - | 9 | 1416 | 5067233 | ||

Scml4
Genomic Map