siRNA Details
| Gene Symbol: Fam19a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252387.1 | NP_001239316.1 | 2 | 1509 | ||
| NM_182807.3 | NP_877959.2 | 3 | 1721 | |||
| GenBank mRNA | AK032483.1 | - | 2 | 591 | ||
| AK139403.1 | - | 2 | 1593 | |||
| AK140763.1 | - | 2 | 1472 | |||
| AK164201.1 | - | 2 | 601 | |||
| AY325121.1 | - | 3 | 408 | |||
| BC027082.1 | - | 3 | 1607 | 5360443 | ||

Fam19a2
Genomic Map