siRNA Details
| Gene Symbol: Sh3pxd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177364.3 | NP_796338.2 | 13 | 1974 | ||
| GenBank mRNA | AB430861.1 | - | 13 | 1896 | ||
| AK089330.1 | - | 13 | 1947 | |||
| AK090119.1 | - | 13 | 1969 | |||
| AK154898.1 | - | 13 | 1932 | |||
| AK171384.1 | - | 13 | 1946 | |||
| BC115711.1 | - | 13 | 1943 | |||
| BC115764.1 | - | 13 | 1943 | |||
| BC141306.1 | - | 13 | 1946 | 9055918 | ||

Sh3pxd2b
Genomic Map