siRNA Details
| Gene Symbol: Ankrd13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172945.2 | NP_766533.2 | 7 | 928 | ||
| GenBank mRNA | BC056928.1 | - | 2 | 157 | ||
| BC151158.1 | - | 7 | 906 | 9088046 | ||
| BC172028.1 | - | 7 | 907 | |||
| CA315262.1 | - | 6 | 614 | |||
| CD578820.1 | - | 1 | 26 | |||

Ankrd13b
Genomic Map