siRNA Details
| Gene Symbol: Phf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174852.3 | NP_777277.2 | 9 | 2452 | ||
| GenBank mRNA | AK050751.1 | - | 3 | 869 | ||
| AK089226.1 | - | 9 | 2515 | |||
| AK129383.1 | - | 6 | 1599 | |||
| AK141796.1 | - | 9 | 2356 | |||
| AK155354.1 | - | 9 | 2493 | |||
| AK170155.1 | - | 9 | 2540 | |||
| AK170187.1 | - | 8 | 1747 | |||
| BC038275.1 | - | 1 | 320 | |||
| BC043080.1 | - | 9 | 2055 | 6400617 | ||

Phf12
Genomic Map