siRNA Details
| Gene Symbol: Zfp652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201609.2 | NP_963903.2 | 1 | 549 | ||
| GenBank mRNA | AK220361.1 | - | 1 | 394 | ||
| BC055752.1 | - | 1 | 549 | 6414140 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp652| Gene Symbol: Zfp652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201609.2 | NP_963903.2 | 1 | 549 | ||
| GenBank mRNA | AK220361.1 | - | 1 | 394 | ||
| BC055752.1 | - | 1 | 549 | 6414140 | ||