siRNA Details
| Gene Symbol: Krt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010661.2 | NP_034791.2 | 1 | 471 | ||
| GenBank mRNA | AK053362.1 | - | 1 | 472 | ||
| AK143182.1 | - | 1 | 470 | |||
| BC131911.1 | - | 1 | 467 | 40130188 | ||
| CK627700.1 | - | 1 | 457 | |||
| U02880.1 | - | 1 | 468 | |||

Krt12
Genomic Map