siRNA Details
| Gene Symbol: Slc26a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178743.3 | NP_848858.2 | 4, 5 | 576 | ||
| GenBank mRNA | AF345196.1 | - | 4, 5 | 576 | ||
| AK078545.1 | - | 4, 5 | 553 | |||
| AK089182.1 | - | 4, 5 | 556 | |||
| BC038604.1 | - | 4, 5 | 569 | |||

Slc26a11
Genomic Map