siRNA Details
| Gene Symbol: Bahcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198423.3 | NP_940815.3 | 26 | 7521 | ||
| GenBank mRNA | AB257855.1 | - | 25 | 7523 | ||
| AK147250.1 | - | 26 | 7521 | |||
| AK147400.1 | - | 18 | 4429 | |||
| AK164229.1 | - | 9 | 1762 | |||
| BC060615.1 | - | 9 | 1793 | |||
| BC072602.1 | - | 15 | 2959 | 6830770 | ||

Bahcc1
Genomic Map