siRNA Details
| Gene Symbol: Bahcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198423.3 | NP_940815.3 | 18 | 5580 | ||
| GenBank mRNA | AB257855.1 | - | 17 | 5582 | ||
| AK147250.1 | - | 18 | 5580 | |||
| AK147400.1 | - | 10 | 2488 | |||
| BC072602.1 | - | 7 | 1018 | 6830770 | ||

Bahcc1
Genomic Map