siRNA Details
| Gene Symbol: Zbtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178744.2 | NP_848859.1 | 2 | 1247 | ||
| GenBank mRNA | AK029762.1 | - | 2 | 995 | ||
| AK035036.1 | - | 2 | 1327 | |||
| AK049397.1 | - | 2 | 1247 | |||
| AK131985.1 | - | 2 | 930 | |||
| AY611626.1 | - | 2 | 1247 | |||
| BC012239.1 | - | 2 | 911 | 4037044 | ||

Zbtb1
Genomic Map