siRNA Details
| Gene Symbol: Gphn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145965.2 | NP_666077.2 | 21, 22 | 2218 | ||
| NM_172952.3 | NP_766540.2 | 22, 23 | 2209 | |||
| GenBank mRNA | AK082353.1 | - | 22, 23 | 2131 | ||
| AK144569.1 | - | 21, 22 | 2221 | |||
| AK220267.1 | - | 3, 4 | 203 | |||
| BC027112.1 | - | 10, 11 | 1005 | |||
| BC127058.1 | - | 21, 22 | 2140 | 40109771 | ||

Gphn
Genomic Map