siRNA Details
| Gene Symbol: Slc38a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178746.4 | NP_848861.1 | 15 | 1711 | ||
| GenBank mRNA | AK043402.1 | - | 15 | 1699 | ||
| AK044735.1 | - | 15 | 1701 | |||
| AK078926.1 | - | 9 | 951 | |||
| BC052361.1 | - | 15 | 1711 | 6705789 | ||

Slc38a9
Genomic Map