siRNA Details
| Gene Symbol: Zswim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252081.1 | NP_001239010.1 | 8 | 1262 | ||
| NM_001252082.1 | NP_001239011.1 | 8 | 1262 | |||
| NM_027996.3 | NP_082272.1 | 8 | 1262 | |||
| GenBank mRNA | AK029263.1 | - | 7 | 718 | ||
| AK053265.1 | - | 8 | 1214 | |||
| AK147398.1 | - | 8 | 1263 | |||
| BC085161.1 | - | 8 | 776 | |||
| BC151046.1 | - | 8 | 1161 | 9087959 | ||
| BC151056.1 | - | 8 | 1161 | 9087969 | ||
| BC151173.1 | - | 8 | 1161 | 9088062 | ||
| BC172103.1 | - | 8 | 1161 | 9054389 | ||

Zswim8
Genomic Map