siRNA Details
| Gene Symbol: Zswim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252081.1 | NP_001239010.1 | 19 | 4005 | ||
| NM_001252082.1 | NP_001239011.1 | 20 | 3924 | |||
| NM_027996.3 | NP_082272.1 | 19 | 4026 | |||
| GenBank mRNA | AB093273.1 | - | 9 | 2102 | ||
| AK147398.1 | - | 19 | 4027 | |||
| BC027155.1 | - | 3 | 478 | |||
| BC030914.1 | - | 3 | 453 | |||
| BC049362.1 | - | 7 | 1077 | |||
| BC059058.1 | - | 10 | 1846 | |||
| BC085161.1 | - | 20 | 3438 | |||
| BC151046.1 | - | 19 | 3925 | 9087959 | ||
| BC151056.1 | - | 19 | 3925 | 9087969 | ||
| BC151173.1 | - | 19 | 3925 | 9088062 | ||
| BC172103.1 | - | 19 | 3901 | 9054389 | ||

Zswim8
Genomic Map