siRNA Details
| Gene Symbol: Gulo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178747.3 | NP_848862.1 | 3 | 383 | ||
| GenBank mRNA | AK077740.1 | - | 3 | 384 | ||
| AK167460.1 | - | 3 | 372 | |||
| AY453064.1 | - | 2 | 220 | |||
| BC019856.1 | - | 3 | 330 | 5123684 | ||
| BC028822.1 | - | 3 | 339 | 5097681 | ||
| BC028828.1 | - | 3 | 342 | 5101228 | ||
| BC034835.1 | - | 2 | 139 | |||

Gulo
Genomic Map