siRNA Details
| Gene Symbol: Nlrc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081280.1 | NP_001074749.1 | 11, 12 | 2964 | ||
| NM_175547.3 | NP_780756.1 | 8, 9 | 887 | |||
| GenBank mRNA | AK079766.1 | - | 8, 9 | 811 | ||
| AK084199.1 | - | 8, 9 | 801 | |||
| AK171971.1 | - | 10, 11 | 3977 | |||
| BC130223.1 | - | 8, 9 | 2353 | |||
| BC152543.1 | - | 8, 9 | 2362 | |||

Nlrc3
Genomic Map