siRNA Details
| Gene Symbol: PSMD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002814.3 | NP_002805.1 | 3 | 449 | ||
| NM_170750.2 | NP_736606.1 | 3 | 449 | |||
| GenBank mRNA | AB009619.1 | - | 3 | 347 | ||
| AK058108.1 | - | 3 | 423 | |||
| AK295996.1 | - | 3 | 343 | |||
| AY057056.1 | - | 3 | 347 | |||
| BC011960.1 | - | 3 | 358 | 3881328 | ||
| BI114640.1 | - | 3 | 456 | |||
| BQ217943.1 | - | 3 | 365 | |||
| BT019689.1 | - | 3 | 325 | |||
| D83197.1 | - | 3 | 423 | |||

PSMD10
Genomic Map