siRNA Details
| Gene Symbol: Atp13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284375.1 | NP_001271304.1 | 26 | 3169 | ||
| NM_175650.4 | NP_783581.2 | 27 | 3304 | |||
| GenBank mRNA | AK083102.1 | - | 28 | 3342 | ||
| AK083121.1 | - | 27 | 3305 | |||
| AK158355.1 | - | 27 | 3124 | |||
| BC115739.1 | - | 26 | 3102 | 40086065 | ||

Atp13a5
Genomic Map