siRNA Details
| Gene Symbol: Lsamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175548.3 | NP_780757.1 | 6 | 948 | ||
| GenBank mRNA | AK044845.1 | - | 6 | 948 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lsamp| Gene Symbol: Lsamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175548.3 | NP_780757.1 | 6 | 948 | ||
| GenBank mRNA | AK044845.1 | - | 6 | 948 | ||