siRNA Details
| Gene Symbol: Dpcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033366.3 | NP_001028538.1 | 2 | 835 | ||
| GenBank mRNA | AK131887.1 | - | 2 | 837 | ||
| AK164929.1 | - | 2 | 836 | |||
| BC150798.1 | - | 2 | 828 | 9087709 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpcr1| Gene Symbol: Dpcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033366.3 | NP_001028538.1 | 2 | 835 | ||
| GenBank mRNA | AK131887.1 | - | 2 | 837 | ||
| AK164929.1 | - | 2 | 836 | |||
| BC150798.1 | - | 2 | 828 | 9087709 | ||