siRNA Details
Gene Symbol: Dpcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033366.3 | NP_001028538.1 | 2 | 1185 | ||
GenBank mRNA | AK131887.1 | - | 2 | 1187 | ||
AK164929.1 | - | 2 | 1186 | |||
BC150798.1 | - | 2 | 1178 | 9087709 |
