siRNA Details
| Gene Symbol: Arhgap28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172964.4 | NP_766552.3 | 4 | 686 | ||
| GenBank mRNA | AK087523.1 | - | 4 | 699 | ||
| AK133720.1 | - | 4 | 528 | |||
| AK141901.1 | - | 4 | 636 | |||
| AK173147.1 | - | 3 | 313 | |||
| BC066788.1 | - | 4 | 676 | 30049271 | ||

Arhgap28
Genomic Map