siRNA Details
| Gene Symbol: Arhgap28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172964.4 | NP_766552.3 | 8 | 1183 | ||
| GenBank mRNA | AK087523.1 | - | 8 | 1196 | ||
| AK133720.1 | - | 8 | 1025 | |||
| AK141901.1 | - | 8 | 1133 | |||
| AK162051.1 | - | 2 | 223 | |||
| AK173147.1 | - | 7 | 810 | |||

Arhgap28
Genomic Map