siRNA Details
| Gene Symbol: Strn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011500.2 | NP_035630.2 | 9 | 1133 | ||
| GenBank mRNA | AJ223777.1 | - | 9 | 1060 | ||
| AK135684.1 | - | 4 | 325 | |||
| AK136032.1 | - | 7 | 782 | |||
| BC090968.1 | - | 9 | 1133 | 30646517 | ||
| BC150727.1 | - | 9 | 1101 | 9087638 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_019148, XM_006239622.

Strn
Genomic Map