siRNA Details
| Gene Symbol: Sap130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172965.2 | NP_766553.1 | 9 | 1172 | ||
| GenBank mRNA | AK034401.1 | - | 9 | 1154 | ||
| AK053675.1 | - | 9 | 1173 | |||
| AK084078.1 | - | 8 | 1320 | |||
| BC063075.1 | - | 6 | 696 | 5721433 | ||
| BC066030.1 | - | 9 | 1184 | |||

Sap130
Genomic Map