siRNA Details
| Gene Symbol: Zfp608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175751.4 | NP_786927.2 | 2 | 875 | ||
| GenBank mRNA | AK047979.1 | - | 2 | 836 | ||
| BC059268.1 | - | 2 | 798 | |||
| BC092219.1 | - | 2 | 608 | 6830664 | ||
| BC151073.1 | - | 1 | 394 | 9087986 | ||
| BC151075.1 | - | 1 | 394 | 9087988 | ||

Zfp608
Genomic Map