siRNA Details
| Gene Symbol: Mgat4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173870.3 | NP_776295.1 | 3 | 553 | ||
| GenBank mRNA | AB053217.1 | - | 2 | 229 | ||
| AK155724.1 | - | 3 | 554 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgat4a| Gene Symbol: Mgat4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173870.3 | NP_776295.1 | 3 | 553 | ||
| GenBank mRNA | AB053217.1 | - | 2 | 229 | ||
| AK155724.1 | - | 3 | 554 | |||