siRNA Details
| Gene Symbol: Nbeal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173444.2 | NP_775620.2 | 25 | 3850 | ||
| GenBank mRNA | AK040951.1 | - | 24 | 3731 | ||
| AK143706.1 | - | 13 | 1759 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nbeal1| Gene Symbol: Nbeal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173444.2 | NP_775620.2 | 25 | 3850 | ||
| GenBank mRNA | AK040951.1 | - | 24 | 3731 | ||
| AK143706.1 | - | 13 | 1759 | |||