siRNA Details
| Gene Symbol: Pask | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080850.2 | NP_543126.2 | 12 | 3486 | ||
| GenBank mRNA | AK028435.1 | - | 12 | 3488 | ||
| AK083528.1 | - | 12 | 3457 | |||
| AK083590.1 | - | 12 | 3452 | |||
| AK129063.1 | - | 12 | 3376 | |||
| AK141220.1 | - | 123 | ||||
| AK172031.1 | - | 12 | 3434 | |||
| BC150821.1 | - | 11 | 3198 | 9087732 | ||

Pask
Genomic Map