siRNA Details
| Gene Symbol: PSME2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002818.2 | NP_002809.2 | 5 | 313 | ||
| XM_006720213.1 | XP_006720276.1 | 5 | 1107 | |||
| GenBank mRNA | AK026580.1 | - | 5 | 334 | ||
| AK225876.1 | - | 5 | 276 | |||
| AK311569.1 | - | 3 | 952 | |||
| AY771595.1 | - | 5 | 262 | |||
| BC004368.1 | - | 5 | 263 | 3640584 | ||
| BC019885.1 | - | 5 | 263 | 4994572 | ||
| BC072025.1 | - | 5 | 262 | 6270762 | ||
| BX161498.1 | - | 5 | 888 | |||
| CR541657.1 | - | 5 | 248 | |||
| CR541743.1 | - | 5 | 248 | |||
| D45248.1 | - | 5 | 313 | |||

PSME2
Genomic Map