siRNA Details
| Gene Symbol: Muc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290786.1 | NP_001277715.1 | 2 | 744 | ||
| NM_172979.3 | NP_766567.1 | 2 | 744 | |||
| GenBank mRNA | AK052840.1 | - | 2 | 631 | ||
| AK142590.1 | - | 2 | 631 | |||
| BC094506.1 | - | 2 | 627 | 5341608 | ||
| BC116796.1 | - | 2 | 623 | 40126115 | ||
| BC119176.1 | - | 2 | 623 | 8733925 | ||

Muc15
Genomic Map