siRNA Details
| Gene Symbol: Slc4a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081162.1 | NP_001074631.1 | 13 | 1553 | ||
| GenBank mRNA | AK052665.1 | - | 8 | 2422 | ||
| AK158115.1 | - | 13 | 1507 | |||
| BC036154.1 | - | 3 | 1131 | |||
| BC111884.1 | - | 12 | 1348 | 40060299 | ||

Slc4a11
Genomic Map