siRNA Details
Gene Symbol: Znf512b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164597.1 | NP_001158069.1 | 5 | 714 | ||
GenBank mRNA | AK129308.1 | - | 5 | 714 | ||
BC056460.1 | - | 3 | 701 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Znf512b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164597.1 | NP_001158069.1 | 5 | 714 | ||
GenBank mRNA | AK129308.1 | - | 5 | 714 | ||
BC056460.1 | - | 3 | 701 |