siRNA Details
| Gene Symbol: Plch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177732.1 | NP_001171203.1 | 11 | 1470 | ||
| NM_001177733.1 | NP_001171204.1 | 11 | 1470 | |||
| NM_183191.3 | NP_899014.2 | 11 | 1470 | |||
| GenBank mRNA | AK173095.1 | - | 3 | 356 | ||
| AY691172.1 | - | 11 | 1470 | |||
| AY691173.1 | - | 11 | 1470 | |||
| AY691174.1 | - | 11 | 1470 | |||
| BC055005.1 | - | 11 | 1547 | 6491217 | ||

Plch1
Genomic Map